Skip to Content
Merck
Triphenylarsine 97%

Triphenylarsine

Triphenylarsine 97%
Synonym(s):
Arsinetriphenyl, Triphenylarsine
Linear Formula:
(C6H5)3As
CAS Number:
Molecular Weight:
306.23

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing