Skip to Content
Merck
Triosmium dodecacarbonyl 98%

Triosmium dodecacarbonyl

Triosmium dodecacarbonyl 98%
Synonym(s):
Osmium carbonyl, tri-Osmium dodecacarbonyl
Linear Formula:
Os3(CO)12
CAS Number:
Molecular Weight:
906.81
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing