Skip to Content
Merck
Triisopropylsilyl chloride 97%

Triisopropylsilyl chloride

Triisopropylsilyl chloride 97%
Synonym(s):
Chlorotriisopropylsilane, TIPSCl, Triisopropylchlorosilane, Triisopropylsilyl chloride
Linear Formula:
[(CH3)2CH]3SiCl
CAS Number:
Molecular Weight:
192.80

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing