Skip to Content
Merck
Trifluoroacetic acid solution (25% solution in water) for protein sequenation

Trifluoroacetic acid solution

Trifluoroacetic acid solution (25% solution in water) for protein sequenation
Synonym(s):
Trifluoroacetic acid solution, TFA
Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02

Products

Product Category
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing