Triamcinolone diacetate
Synonym(s):
(11β,16α)-16,21-Bis(acetyloxy)-9-fluoro-11,17-dihydroxypregna-1,4-diene-3,20-dione, 9α-Fluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,21-diacetate, 9α-Fluoro-16α-hydroxyprednisolone 16,21-diacetate
Empirical Formula (Hill Notation):
C25H31FO8
CAS Number:
Molecular Weight:
478.51
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing