Skip to Content
Merck
vibrant-m

Tranylcypromine Related Compound A

vibrant-m
Synonym(s):
cis-Tranylcypromine hydrochloride, (±)-cis-2-Phenylcyclopropanamine hydrochloride
Empirical Formula (Hill Notation):
C9H11N · HCl
CAS Number:
Molecular Weight:
169.65

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade