Skip to Content
Merck
trans,trans-Muconic acid-13C6 ≥99 atom % 13C, ≥98% (CP)

trans,trans-Muconic acid-13C6

trans,trans-Muconic acid-13C6 ≥99 atom % 13C, ≥98% (CP)
Synonym(s):
trans,trans-2,4-Hexadienedioic acid-13C6
Empirical Formula (Hill Notation):
13C6H6O4
Molecular Weight:
148.07

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Isotope
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing