Skip to Content
Merck
trans-2-Phenylvinylboronic acid 97%

trans-2-Phenylvinylboronic acid

trans-2-Phenylvinylboronic acid 97%
Synonym(s):
(E)-2-phenyl-Etheneboronic acid, (E)-Phenylethenylboronic acid, (E)-Styreneboronic acid, (E)-Styrylboronic acid, trans-(2-Phenylethenyl)boronic acid, trans-Phenylvinyl boronic acid
Linear Formula:
C6H5CH=CHB(OH)2
CAS Number:
Molecular Weight:
147.97

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing