Skip to Content
Merck
trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile matrix substance for MALDI-MS, ≥99.0% (HPLC)

trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile

trans-2-[3-(4-tert-Butylphenyl)-2-methyl-2-propenylidene]malononitrile matrix substance for MALDI-MS, ≥99.0% (HPLC)
Synonym(s):
DCTB
Empirical Formula (Hill Notation):
C17H18N2
CAS Number:
Molecular Weight:
250.34

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Technique