trans-1-Penten-1-ylboronic acid pinacol ester
Synonym(s):
E-2-(1-Pentenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, trans-1-Pentenylboronic acid pinacol ester
Empirical Formula (Hill Notation):
C11H21BO2
CAS Number:
Molecular Weight:
196.09
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing