trans-1,3-Diphenyl-2-propen-1-ol
Empirical Formula (Hill Notation):
C15H14O
CAS Number:
Molecular Weight:
210.27
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing