![Timolol impurity F European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/231/420/21af5fdf-d3f6-4d5c-b02c-042c4ed5ef85/640/21af5fdf-d3f6-4d5c-b02c-042c4ed5ef85.png)
Timolol impurity F
![Timolol impurity F European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/231/420/21af5fdf-d3f6-4d5c-b02c-042c4ed5ef85/640/21af5fdf-d3f6-4d5c-b02c-042c4ed5ef85.png)
Synonym(s):
3-Chloro-4-morpholin-4-yl-1,2,5-thiadiazole, 4-(4-Chloro-1,2,5-thiadiazol-3-yl)morpholine
Empirical Formula (Hill Notation):
C6H8ClN3OS
Products
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing