
Ticarcillin impurity A

Synonym(s):
(2S,5R,6R)-3,3-Dimethyl-7-oxo-6-[[(thiophen-3-yl)acetyl]amino]-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, Decarboxyticarcillin
Empirical Formula (Hill Notation):
C14H16N2O4S2
CAS Number:
Molecular Weight:
340.42
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing