Skip to Content
Merck
tetra-Sodium diphosphate decahydrate for analysis EMSURE® ACS,Reag. Ph Eur

tetra-Sodium diphosphate decahydrate

tetra-Sodium diphosphate decahydrate for analysis EMSURE® ACS,Reag. Ph Eur
Synonym(s):
tetra-Sodium diphosphate decahydrate
Empirical Formula (Hill Notation):
Na4O7P2 · 10H2O
CAS Number:
Molecular Weight:
446.06

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing