Tetraethylenepentamine
Synonym(s):
TEPA, Tetrene, Tetraethylene pentamine
Linear Formula:
(NH2CH2CH2NHCH2CH2)2NH
CAS Number:
Molecular Weight:
189.30
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing