Skip to Content
Merck
Tetra(ethylene glycol) diacrylate technical grade, contains 100-150 ppm HQ as inhibitor, 150-200 ppm MEHQ as inhibitor

Tetra(ethylene glycol) diacrylate

Tetra(ethylene glycol) diacrylate technical grade, contains 100-150 ppm HQ as inhibitor, 150-200 ppm MEHQ as inhibitor
Synonym(s):
TTEGDA
Linear Formula:
(H2C=CHCO2CH2CH2OCH2CH2)2O
CAS Number:
Molecular Weight:
302.32
EC Number:

Products

Product Category
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type
Special Grade