![Terbinafine Related Compound D certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/574/467/cbc62ba2-1339-4d06-8582-c28f415cad1a/640/cbc62ba2-1339-4d06-8582-c28f415cad1a.jpg)
Terbinafine Related Compound D
![Terbinafine Related Compound D certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/574/467/cbc62ba2-1339-4d06-8582-c28f415cad1a/640/cbc62ba2-1339-4d06-8582-c28f415cad1a.jpg)
Synonym(s):
4-Methylterbinafine hydrochloride, (2E)-N,6,6-Trimethyl-N-[(4-methylnaphthalen-1-yl)methyl]-hept-2-en-4-yn-1-amine hydrochloride, N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N,4-dimethyl-1-naphthalenemethanamine hydrochloride
Empirical Formula (Hill Notation):
C22H27N · HCl
CAS Number:
Molecular Weight:
341.92
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2