![Terbinafine Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/354/894/97f1e476-dba5-4eb8-9e1a-9ab54511d510/640/97f1e476-dba5-4eb8-9e1a-9ab54511d510.png)
Terbinafine Related Compound A
![Terbinafine Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/354/894/97f1e476-dba5-4eb8-9e1a-9ab54511d510/640/97f1e476-dba5-4eb8-9e1a-9ab54511d510.png)
Synonym(s):
N-Methyl-1-naphthalenemethylamine hydrochloride
Linear Formula:
C10H7CH2NHCH3 · HCl
CAS Number:
Molecular Weight:
207.70
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2