Skip to Content
Merck
Tacrolimus Pharmaceutical Secondary Standard; Certified Reference Material

Tacrolimus

Tacrolimus Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
FK-506 monohydrate, Tacrolimus
Empirical Formula (Hill Notation):
C44H69NO12 · H2O
CAS Number:
Molecular Weight:
822.03

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique