![SUPELCOSIL™ LC-SCX Supelguard Cartridge 5 μm particle size, L × I.D. 2 cm × 4 mm](/deepweb/assets/sigmaaldrich/product/images/172/317/d383d954-5c59-4fc6-810d-1f3ef910f6c4/640/d383d954-5c59-4fc6-810d-1f3ef910f6c4.jpg)
SUPELCOSIL™ LC-SCX (5 µm) HPLC Columns
![SUPELCOSIL™ LC-SCX Supelguard Cartridge 5 μm particle size, L × I.D. 2 cm × 4 mm](/deepweb/assets/sigmaaldrich/product/images/172/317/d383d954-5c59-4fc6-810d-1f3ef910f6c4/640/d383d954-5c59-4fc6-810d-1f3ef910f6c4.jpg)
Products
Product Category
Brand
pH Value
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing