![Sucrose Optical Rotation NIST® SRM® 17g](/deepweb/assets/sigmaaldrich/product/structures/672/961/1cd4ed88-15e4-4ea3-8920-2c857eb9bb72/640/1cd4ed88-15e4-4ea3-8920-2c857eb9bb72.png)
Sucrose Optical Rotation
![Sucrose Optical Rotation NIST® SRM® 17g](/deepweb/assets/sigmaaldrich/product/structures/672/961/1cd4ed88-15e4-4ea3-8920-2c857eb9bb72/640/1cd4ed88-15e4-4ea3-8920-2c857eb9bb72.png)
Synonym(s):
Sucrose, α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar
Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Optical Isomer
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing