Skip to Content
BE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
GF95244082
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 50mm disks, 0.1mm thickness, annealed
Expand
View Pricing
GF20115706
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 150 x 150mm, 0.1mm thickness, annealed
Expand
View Pricing
GF95961954
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 25mm disks, 0.1mm thickness, annealed
Expand
View Pricing
GF68800080
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 6mm disks, 0.1mm thickness, annealed
Expand
View Pricing
GF03572489
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 50 x 50mm, 0.1mm thickness, annealed
Expand
View Pricing
GF53022541
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
powder, 45micron max. particle size, atomized, 500g, 99.999%
Expand
View Pricing
GF00010449
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 8mm disks, 0.1mm thickness, annealed
Expand
View Pricing
GF56550405
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 4mm disks, 0.1mm thickness, annealed
Expand
View Pricing
GF46560922
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 100 x 100mm, 0.1mm thickness, annealed
Expand
View Pricing
GF21628571
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 25 x 25mm, 0.1mm thickness, annealed
Expand
View Pricing
GF63167698
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 15mm disks, 0.1mm thickness, annealed
Expand
View Pricing
GF36444999
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
powder, 45micron max. particle size, atomized, 100g, 99.999%
Expand
View Pricing
GF01305346
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 10mm disks, 0.1mm thickness, annealed
Expand
View Pricing
GF14463367
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 300 x 300mm, 0.1mm thickness, annealed
Expand
View Pricing
GF93447119
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 150 x 150mm, 2.0mm thickness, annealed
Expand
View Pricing
GF55624181
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
powder, 45micron max. particle size, atomized, 200g, 99.999%
Expand
View Pricing
GF19230566
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 300 x 300mm, 2.0mm thickness, annealed
Expand
View Pricing
GF07920413
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 1m coil, 0.1mm thickness, annealed
Expand
View Pricing
GF90052400
Stainless Steel - AISI 347 alloy, FeCr18Ni10Nb,
foil, 2m coil, 0.1mm thickness, annealed
Expand
View Pricing