![(S)-(−)-Perillyl alcohol ≥95%, FG](/deepweb/assets/sigmaaldrich/product/structures/282/498/b9a8d961-67ad-49bd-be7e-110784caacd6/640/b9a8d961-67ad-49bd-be7e-110784caacd6.png)
(S)-(−)-Perillyl alcohol
![(S)-(−)-Perillyl alcohol ≥95%, FG](/deepweb/assets/sigmaaldrich/product/structures/282/498/b9a8d961-67ad-49bd-be7e-110784caacd6/640/b9a8d961-67ad-49bd-be7e-110784caacd6.png)
Synonym(s):
p-Mentha-1,8-diene-7-ol
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing