Skip to Content
Merck
Sodium thiocyanate free-flowing, Redi-Dri™, ACS reagent, ≥98.0%

Sodium thiocyanate

Sodium thiocyanate free-flowing, Redi-Dri™, ACS reagent, ≥98.0%
Synonym(s):
Sodium isothiocyanate, Sodium rhodanate, Sodium rhodanide, Sodium sulfocyanate, Sodium thiocyanate
Linear Formula:
NaSCN
CAS Number:
Molecular Weight:
81.07

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Special Grade