![Sodium Oxalate Reductometric Standard NIST®SRM®](/deepweb/assets/sigmaaldrich/product/structures/175/213/ac7d7ee8-4e7b-4f24-a419-770316329cd4/640/ac7d7ee8-4e7b-4f24-a419-770316329cd4.png)
Sodium Oxalate Reductometric Standard
![Sodium Oxalate Reductometric Standard NIST®SRM®](/deepweb/assets/sigmaaldrich/product/structures/175/213/ac7d7ee8-4e7b-4f24-a419-770316329cd4/640/ac7d7ee8-4e7b-4f24-a419-770316329cd4.png)
Synonym(s):
Sodium oxalate, Ethanedioic acid sodium salt, Oxalic acid disodium salt
Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing