Skip to Content
Merck
Sodium nitroferricyanide(III) dihydrate ACS reagent, ≥99%

Sodium nitroferricyanide(III) dihydrate

Sodium nitroferricyanide(III) dihydrate ACS reagent, ≥99%
Synonym(s):
Nitroprusside sodium, SNP, Sodium nitroprusside, Sodium pentacyanonitrosylferrate
Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
CAS Number:
Molecular Weight:
297.95
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing