Sodium 1,1-difluoroethanesulfinate
Synonym(s):
Baran DFES Reagent, Baran Difluoroethylation Reagent, DFES-Na Reagent, Sodium 1,1-difluoroethylsulfinate
Empirical Formula (Hill Notation):
C2H3F2NaO2S
CAS Number:
Molecular Weight:
152.10
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing