Skip to Content
Merck
(S)-(−)-N,N-Dimethyl-1-ferrocenylethylamine ≥98.0% (T)

(S)-(−)-N,N-Dimethyl-1-ferrocenylethylamine

(S)-(−)-N,N-Dimethyl-1-ferrocenylethylamine ≥98.0% (T)
Synonym(s):
(S)-(−)-[1-(Dimethylamino)ethyl]ferrocene
Empirical Formula (Hill Notation):
C14H19FeN
CAS Number:
Molecular Weight:
257.15

Products

Brand
Boiling Point (°C)
Color
Formula Weight
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing