Skip to Content
Merck
S-Nitroso-N-acetyl-DL-penicillamine ≥97%, powder

S-Nitroso-N-acetyl-DL-penicillamine

S-Nitroso-N-acetyl-DL-penicillamine ≥97%, powder
Synonym(s):
N-Acetyl-3-(nitrosothio)-DL-valine, S-Nitroso-N-acetylpenicillamine, SNAP
Empirical Formula (Hill Notation):
C7H12N2O4S
CAS Number:
Molecular Weight:
220.25

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing