Sinomenine hydrochloride
Synonym(s):
(9a,13a,14a)-7,8-Didehydro-4-hydroxy-3,7-dimethoxy-17-methyl-morphinan-6-one hydrochloride, Cucoline hydrochloride, Kukolin hydrochloride
Empirical Formula (Hill Notation):
C19H23NO4 · HCl
CAS Number:
Molecular Weight:
365.85
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing