![Sennidin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/291/444/1d711999-31d9-4e57-9236-c43697ef5a8d/640/1d711999-31d9-4e57-9236-c43697ef5a8d.png)
Sennidin A
![Sennidin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/291/444/1d711999-31d9-4e57-9236-c43697ef5a8d/640/1d711999-31d9-4e57-9236-c43697ef5a8d.png)
Synonym(s):
(9R)-9-[(9R)-2-carboxy-4,5-dihydroxy-10-oxo-9H-anthracen-9-yl]-4,5-dihydroxy-10-oxo-9H-anthracene-2-carboxylic acid, Dihydroxydianthrone
Empirical Formula (Hill Notation):
C30H18O10
CAS Number:
Molecular Weight:
538.46
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing