(S)-(+)-2-Amino-4-bromobutyric acid hydrobromide
Linear Formula:
BrCH2CH2CH(NH2)CO2H · HBr
CAS Number:
Molecular Weight:
262.93
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing