![Ritodrine hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/201/408/4726be7f-69ec-4ae4-8536-8a7f2a91757b/640/4726be7f-69ec-4ae4-8536-8a7f2a91757b.png)
Ritodrine hydrochloride
![Ritodrine hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/201/408/4726be7f-69ec-4ae4-8536-8a7f2a91757b/640/4726be7f-69ec-4ae4-8536-8a7f2a91757b.png)
Synonym(s):
N-(p-Hydroxyphenethyl)-4-hydroxynorephedrine hydrochloride, Ritodrine hydrochloride
Empirical Formula (Hill Notation):
C17H21NO3 · HCl
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing