(R)-(−)-Glycidyl butyrate
Synonym(s):
(R)-(−)-Oxirane-2-methanol butyrate
Empirical Formula (Hill Notation):
C7H12O3
CAS Number:
Molecular Weight:
144.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing