Retronecine
Synonym(s):
(1R,7aR)-2,3,5,7a-Tetrahydro-1-hydroxy-1H-pyrrolizine-7-methanol
Empirical Formula (Hill Notation):
C8H13NO2
CAS Number:
Molecular Weight:
155.19
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing