![Resorufin sodium salt redox indicator](/deepweb/assets/sigmaaldrich/product/structures/715/621/9a61d4ab-bd50-469d-9bf4-4030b2c2bc4d/640/9a61d4ab-bd50-469d-9bf4-4030b2c2bc4d.png)
Resorufin sodium salt
![Resorufin sodium salt redox indicator](/deepweb/assets/sigmaaldrich/product/structures/715/621/9a61d4ab-bd50-469d-9bf4-4030b2c2bc4d/640/9a61d4ab-bd50-469d-9bf4-4030b2c2bc4d.png)
Synonym(s):
7-Hydroxy-3H-phenoxazin-3-one sodium salt
Empirical Formula (Hill Notation):
C12H6NNaO3
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing