![(R)-(+)-α,4-Dimethylbenzylamine ChiPros®, produced by BASF, 99%](/deepweb/assets/sigmaaldrich/product/structures/890/132/efe43e44-4138-49cf-b08e-8aa9d0a3b391/640/efe43e44-4138-49cf-b08e-8aa9d0a3b391.png)
(R)-(+)-α,4-Dimethylbenzylamine
![(R)-(+)-α,4-Dimethylbenzylamine ChiPros®, produced by BASF, 99%](/deepweb/assets/sigmaaldrich/product/structures/890/132/efe43e44-4138-49cf-b08e-8aa9d0a3b391/640/efe43e44-4138-49cf-b08e-8aa9d0a3b391.png)
Linear Formula:
CH3C6H4CH(CH3)NH2
CAS Number:
Molecular Weight:
135.21
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing