Rac Inhibitor III
Synonym(s):
EHop-016, N4-(9-Ethyl-9H-carbazol-3-yl)-N2-[3-(4-morpholinyl)propyl]-2,4-pyrimidinediamine
Empirical Formula (Hill Notation):
C25H30N6O
CAS Number:
Molecular Weight:
430.55
Products
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing