
Pyruvonitrile

Synonym(s):
2-Oxopropionitrile, Acetyl cyanide
Linear Formula:
CH3COCN
CAS Number:
Molecular Weight:
69.06
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1