Pyridostigmine bromide
Synonym(s):
3-(Dimethylaminocarbonyloxy)-1-methylpyridinium bromide, Pyridostigmine bromide
Empirical Formula (Hill Notation):
C9H13BrN2O2
CAS Number:
Molecular Weight:
261.12
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing