Proteose Peptone (vegetable)
Synonym(s):
Peptone from vegetable
CAS Number:
EC Number:
Products
Product Category
Biological Source
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing