Skip to Content
Merck
Propionaldehyde-DNPH solution 1000 μg/mL in acetonitrile (as the aldehyde), ampule of 1 mL

Propionaldehyde-DNPH solution

Propionaldehyde-DNPH solution 1000 μg/mL in acetonitrile (as the aldehyde), ampule of 1 mL
Synonym(s):
Propionaldehyde-2,4-dinitrophenylhydrazone solution
CAS Number:

Products

Brand
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1