Skip to Content
Merck
Pregabalin solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Pregabalin solution

Pregabalin solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
Pregabalin solution
Empirical Formula (Hill Notation):
C8H17NO2
CAS Number:
Molecular Weight:
159.23

Products

Brand
Feature
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique