Skip to Content
Merck
Praziquantel Related Compound C pharmaceutical secondary standard, certified reference material

Praziquantel Related Compound C

Praziquantel Related Compound C pharmaceutical secondary standard, certified reference material
Synonym(s):
2-(N-Formylhexahydrohippuroyl)-1,2,3,4-tetrahydroisoquinolin-1-one
Empirical Formula (Hill Notation):
C19H22N2O4
CAS Number:
Molecular Weight:
342.39

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade