Skip to Content
Merck
Potassium Sucrose Octasulfate certified reference material, pharmaceutical secondary standard

Potassium Sucrose Octasulfate

Potassium Sucrose Octasulfate certified reference material, pharmaceutical secondary standard
Synonym(s):
Sucrose octasulfate potassium salt
Empirical Formula (Hill Notation):
C12H14K8O35S8
CAS Number:
Molecular Weight:
1287.52

Products

Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1