Poly[(R)-3-hydroxybutyric acid-co-(R)-3-hydroxyvaleric acid]
Synonym(s):
Biopol, Copolymer of (R)-3-hydroxybutyric acid and (R)-3-hydroxyvaleric acid
CAS Number:
Products
Product Category
Brand
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing