![Poly[(R)-3-hydroxybutyric acid] natural origin](/deepweb/assets/sigmaaldrich/product/structures/129/476/7d1c924b-f644-4889-a2d6-d7a923ce382c/640/7d1c924b-f644-4889-a2d6-d7a923ce382c.png)
Poly[(R)-3-hydroxybutyric acid]
![Poly[(R)-3-hydroxybutyric acid] natural origin](/deepweb/assets/sigmaaldrich/product/structures/129/476/7d1c924b-f644-4889-a2d6-d7a923ce382c/640/7d1c924b-f644-4889-a2d6-d7a923ce382c.png)
Synonym(s):
(R)-3-Hydroxybutyric acid polymerized, PHB
Linear Formula:
[COCH2CH(CH3)O]n
CAS Number:
Products
Brand
Quality Segment
Polymer Composition
Polymer Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing