Skip to Content
Merck
Poly(ethylene glycol) dimethacrylate average Mn 4,000, contains MEHQ as inhibitor

Poly(ethylene glycol) dimethacrylate

Poly(ethylene glycol) dimethacrylate average Mn 4,000, contains MEHQ as inhibitor
Synonym(s):
PEG dimethacrylate
Linear Formula:
C3H5C(O)(OCH2CH2)nOC(O)C3H5
CAS Number:

Products

Brand
Boiling Point (°C)
Color
Physical Form
Purity
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type
Showing 1-9 of 9
Compare
Product Number
Description
Pricing