![Poly(ethylene glycol)-block-poly(ε−caprolactone) methyl ether PEG average Mn ~5,000, PCL average Mn ~32,000](/deepweb/assets/sigmaaldrich/product/structures/234/569/5c0c278a-7c8d-4438-8448-13676bfe3f28/640/5c0c278a-7c8d-4438-8448-13676bfe3f28.png)
Poly(ethylene glycol)-block-poly(ε−caprolactone) methyl ether
![Poly(ethylene glycol)-block-poly(ε−caprolactone) methyl ether PEG average Mn ~5,000, PCL average Mn ~32,000](/deepweb/assets/sigmaaldrich/product/structures/234/569/5c0c278a-7c8d-4438-8448-13676bfe3f28/640/5c0c278a-7c8d-4438-8448-13676bfe3f28.png)
Synonym(s):
Poly(ethylene glycol) methyl ether-block-poly(ε−caprolactone)
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing