
Phytosphingosine hydrochloride

Synonym(s):
4-Hydroxysphinganine, D-Ribo-1,3,4-trihydroxy-2-aminooctadecane, D-Ribo-2-aminooctadecane-1,3,4-triol, PHS
Empirical Formula (Hill Notation):
C18H39NO3 · HCl
CAS Number:
Molecular Weight:
353.97
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing